2,5-Difluoropyridine ≥98%

Supplier: Matrix Scientific

031254-1G 031254-5G 031254-25G
101831-320EA 24.62 USD
101831-320 101831-322 101831-324
2,5-Difluoropyridine ≥98%
2,5-Difluoropyridine

Hazard code(s): Flammable

Formula: C₅H₃F₂N
MDL Number: MFCD04114191
CAS Number: 84476-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR