Indole-6-carbaldehyd

Supplier: Matrix Scientific

Synonyms: Indole-6-carboxaldehyde, 1H-Indole-6-carbaldehyde

031026-5G 031026-25G
101831-228EA 55.38 USD
101831-228 101831-230
Indole-6-carbaldehyd
Indole-6-carbaldehyd
Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD02179596
CAS Number: 1196-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR