6-Bromoisatin ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-1H-indole-2,3-dione, 6-Bromoindoline-2,3-dione

030121-5G 030121-25G
101829-514EA 36.92 USD
101829-514 101829-516
6-Bromoisatin ≥95%
6-Bromoisatin

Hazard code(s): Irritant, Refrigerate

Formula: C₈H₄BrNO₂
MW: 226.03 g/mol
MDL Number: MFCD01631138
CAS Number: 6326-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR