N,N-Dimethylglycine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid

027373-25G 027373-100G
101824-050EA 29.23 USD
101824-050 101824-052
N,N-Dimethylglycine ≥98%
N,N-Dimethylglycine

Hazard code(s): Irritant

Formula: C₄H₉NO₂
MW: 103.12 g/mol
MDL Number: MFCD00004283
CAS Number: 1118-68-9
EINECS: 214-267-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR