2-Amino-6-chlorobenzothiazole ≥98%

Supplier: Matrix Scientific

027268-5G 027268-25G
101823-852EA 15.38 USD
101823-852 101823-854
2-Amino-6-chlorobenzothiazole ≥98%
2-Amino-6-chlorobenzothiazole

Hazard code(s): Irritant

Formula: C₇H₅ClN₂S
Boiling Pt: 344 °C (1013 hPa)
Melting Pt: 160…163 °C
MDL Number: MFCD00053557
CAS Number: 95-24-9
EINECS: 202-402-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR