5-Methylisoxazole-3-carboxylic acid

Supplier: Matrix Scientific

027169-5G 027169-25G
101823-644EA 69.23 USD
101823-644 101823-646
5-Methylisoxazole-3-carboxylic acid
5-Methylisoxazole-3-carboxylic acid
Formula: C₅H₅NO₃
MW: 127.1 g/mol
MDL Number: MFCD01318162
CAS Number: 3405-77-4
EINECS: 222-289-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR