5-Amino-1,2,4-thiadiazole ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: 1,2,4-Thiadiazol-5-amine

027101-1G 027101-5G
101823-492EA 89.23 USD
101823-492 101823-494
5-Amino-1,2,4-thiadiazole ≥95% for synthesis
5-Amino-1,2,4-thiadiazole

Hazard code(s): Irritant

Formula: C₂H₃N₃S
Melting Pt: 112…114 °C
MDL Number: MFCD01102171
CAS Number: 7552-07-0

Order Now

Specification Test Results

Melting point 112-114°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR