2-Amino-3-fluoropyridine ≥97%
Supplier: Matrix Scientific
026754-1G
026754-5G
101822-804EA
21.54
USD
101822-804
101822-806
2-Amino-3-fluoropyridine ≥97%
2-Amino-3-fluoropyridine
Hazard code(s): Toxic
Formula:
C₅H₅FN₂ Melting Pt: 39…43 °C |
MDL Number:
MFCD04114135 CAS Number: 21717-95-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...