3-Fluoro-2-nitropyridine ≥97%
Supplier: Matrix Scientific
026753-1G
026753-5G
101822-800EA
29.23
USD
101822-800
101822-802
3-Fluoro-2-nitropyridine ≥97%
3-Fluoro-2-nitropyridine
Hazard code(s): Irritant, Store in Freezer
Formula:
C₅H₃FN₂O₂ Melting Pt: 30…35 °C |
MDL Number:
MFCD04114127 CAS Number: 54231-35-5 EINECS: 207-260-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...