Piperidin-3-ol ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3-Piperidinol, 3-Hydroxypiperidine

026734-5G 026734-25G 026734-100G
101822-760EA 29.23 USD
101822-760 101822-762 101822-764
Piperidin-3-ol ≥98%
Piperidin-3-ol

Hazard code(s): Corrosive, Air Sensitive, Hygroscopic

Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 67…69 °C (2mm)
Melting Pt: 56…60 °C
MDL Number: MFCD00014591
CAS Number: 6859-99-0
EINECS: 229-957-4
UN: 3263
ADR: 8,III

Order Now

Specification Test Results

Boiling point 67-69°C/2mm
Melting point 56-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR