2-Amino-3,5-difluoropyridine ≥98%
Supplier: Matrix Scientific
Synonyms:
3,5-Difluoro-2-pyridylamine, 3,5-Difluoropyridin-2-amine
026729-1G
026729-5G
101822-740EA
30.77
USD
101822-740
101822-742
2-Amino-3,5-difluoropyridine ≥98%
2-Amino-3,5-difluoropyridine
Hazard code(s): Irritant
Formula:
C₅H₄F₂N₂ Melting Pt: 55…57 °C |
MDL Number:
MFCD03788760 CAS Number: 732306-31-9 UN: 2811 ADR: 6.1,III |
Specification Test Results
Melting point | 55-57°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...