3-Bromo-2-methoxy-5-nitropyridine ≥98% for synthesis

Supplier: Matrix Scientific

026728-5G 026728-25G
101822-736EA 60 USD
101822-736 101822-738
3-Bromo-2-methoxy-5-nitropyridine ≥98% for synthesis
3-Bromo-2-methoxy-5-nitropyridine

Hazard code(s): Irritant

Formula: C₆H₅BrN₂O₃
MW: 233.02
MDL Number: MFCD06659517
CAS Number: 15862-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR