3-Bromo-2-fluoropyridine ≥98%

Supplier: Matrix Scientific

026722-5G 026722-25G
101822-726EA 27.69 USD
101822-726 102663-652
3-Bromo-2-fluoropyridine ≥98%
3-Bromo-2-fluoropyridine

Hazard code(s): Toxic

Formula: C₅H₃BrFN
MDL Number: MFCD04112496
CAS Number: 36178-05-9
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR