2-Fluoro-5-iodopyridine ≥98% for synthesis

Supplier: Matrix Scientific

026721-5G 026721-25G
101822-720EA 41.54 USD
101822-720 101822-722
2-Fluoro-5-iodopyridine ≥98% for synthesis
2-Fluoro-5-iodopyridine

Hazard code(s): Irritant

Formula: C₅H₃FIN
Melting Pt: 31…33 °C
MDL Number: MFCD03095301
CAS Number: 171197-80-1

Order Now

Specification Test Results

Melting point 31-33°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR