2-Bromo-6-chlorobenzoic acid ≥98%

Supplier: Matrix Scientific

026302-5G 026302-25G 026302-100G
101822-038EA 18.46 USD
101822-038 101822-040 101822-042
2-Bromo-6-chlorobenzoic acid ≥98%
2-Bromo-6-chlorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Melting Pt: 148…150 °C
MDL Number: MFCD00672929
CAS Number: 93224-85-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 148-150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR