2-Bromo-5-nitro-6-picoline ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-2-methyl-3-nitropyridine, 2-Bromo-6-methyl-5-nitropyridine

026295-5G 026295-25G
101822-012EA 40 USD
101822-012 101822-014
2-Bromo-5-nitro-6-picoline ≥98%
2-Bromo-5-nitro-6-picoline

Hazard code(s): Irritant

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
MDL Number: MFCD03095219
CAS Number: 22282-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR