4-Chloropicolinic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Chloropyridine-2-carboxylic acid

026120-5G 026120-25G
101821-642EA 46.15 USD
101821-642 101821-644
4-Chloropicolinic acid ≥98%
4-Chloropicolinic acid

Hazard code(s): Irritant

Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
MDL Number: MFCD00191400
CAS Number: 5470-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR