3-Formyl-2-methoxypyridine

Supplier: Matrix Scientific

Synonyms: 2-Methoxynicotinaldehyde, 2-Methoxy-3-pyridinecarboxaldehyde, 2-Methoxy-pyridine-3-carbaldehyde, 2-Methoxypyridine-3-carboxaldehyde

026108-500MG 026108-1G 026108-5G
101821-618EA 29.23 USD
101821-618 102551-078 102551-080
3-Formyl-2-methoxypyridine
3-Formyl-2-methoxypyridine
Formula: C₇H₇NO₂
MW: 137.14 g/mol
MDL Number: MFCD04115112
CAS Number: 71255-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR