3,4-Dimethoxycinnamic acid

Supplier: Matrix Scientific

Synonyms: 3,4-Dimethoxycinnamic Acid

025954-5G 025954-25G
101821-314EA 18.46 USD
101821-314 101821-316
3,4-Dimethoxycinnamic acid
3,4-Dimethoxycinnamic acid

Hazard code(s): Irritant

Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 198 - 202 °C
MDL Number: MFCD00004387
CAS Number: 2316-26-9
EINECS: 219-025-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR