4-Bromo-2-methoxybenzoic acid ≥98%

Supplier: Matrix Scientific

024997-5G 024997-25G
101819-370EA 41.54 USD
101819-370 101819-372
4-Bromo-2-methoxybenzoic acid ≥98%
4-Bromo-2-methoxybenzoic acid

Hazard code(s): Irritant

Formula: C₈H₇BrO₃
MW: 231.05
MDL Number: MFCD07780665
CAS Number: 72135-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR