2,3-Difluoro-5-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

024995-1G 024995-5G 024995-25G
101819-360EA 30.77 USD
101819-360 101819-362 101819-364
2,3-Difluoro-5-(trifluoromethyl)pyridine ≥98%
2,3-Difluoro-5-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₆H₂F₅N
MW: 183.08 g/mol
Boiling Pt: 104 °C
Density: 1.47 g/cm³
MDL Number: MFCD04972837
CAS Number: 89402-42-6

Order Now

Specification Test Results

Boiling point 104°C
Density 1.47

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR