3-Bromo-4-chloropyridine ≥98%

Supplier: Matrix Scientific

024993-5G 024993-25G
101819-356EA 40 USD
101819-356 101819-358
3-Bromo-4-chloropyridine ≥98%
3-Bromo-4-chloropyridine

Hazard code(s): Irritant, Refrigerate

Formula: C₅H₃BrClN
Boiling Pt: 93…95 °C (5mm)
Melting Pt: 31…34 °C
Density: 1.19 g/cm³
MDL Number: MFCD00234008
CAS Number: 36953-42-1

Order Now

Specification Test Results

Boiling point 93-95°C/5mm
Melting point 31-34°C
Density 1.19
ND20 1.453


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR