2-Fluoro-4-(trifluoromethyl)pyridine ≥97%
Supplier: Matrix Scientific
024840-1G
024840-5G
101819-038EA
21.54
USD
101819-038
101819-040
2-Fluoro-4-(trifluoromethyl)pyridine ≥97%
2-Fluoro-4-(trifluoromethyl)pyridine
Hazard code(s): Irritant
Formula:
C₆H₃F₄N MW: 165.09 g/mol Boiling Pt: 110 °C |
MDL Number:
MFCD09879262 CAS Number: 118078-66-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...