7-Bromo-1-tetralone ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 7-Bromo-3,4-dihydronaphthalen-1(2H)-one

024830-1G 024830-5G 024830-25G
101819-012EA 27.69 USD
101819-012 101819-014 101819-016
7-Bromo-1-tetralone ≥97% for synthesis
7-Bromo-1-tetralone

Hazard code(s): Irritant

Formula: C₁₀H₉BrO
Melting Pt: 73…77 °C
MDL Number: MFCD02179287
CAS Number: 32281-97-3

Order Now

Specification Test Results

Melting point 73-77°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR