5-Hydroxy-2-(trifluoromethyl)pyridine ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 6-(Trifluoromethyl)pyridin-3-ol, 2-(Trifluoromethyl)pyridin-5-ol

024816-1G 024816-5G
101818-976EA 44.62 USD
101818-976 101818-978
5-Hydroxy-2-(trifluoromethyl)pyridine ≥98% for synthesis
5-Hydroxy-2-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₆H₄F₃NO
Melting Pt: 174…176 °C
MDL Number: MFCD06801325
CAS Number: 216766-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR