2-Bromo-5-nitro-3-picoline ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-3-methyl-5-nitropyridine

024814-5G 024814-25G
101818-968EA 44.62 USD
101818-968 101818-970
2-Bromo-5-nitro-3-picoline ≥98%
2-Bromo-5-nitro-3-picoline

Hazard code(s): Irritant

Formula: C₆H₅BrN₂O₂
MW: 217.02 g/mol
Melting Pt: 57…58 °C
MDL Number: MFCD03095065
CAS Number: 23132-21-0

Order Now

Specification Test Results

Melting point 57-58°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR