2-Amino-5-fluoro-4-methylpyridine ≥95% for synthesis

Supplier: Matrix Scientific

024812-1G 024812-5G
101818-958EA 89.23 USD
101818-958 101818-960
2-Amino-5-fluoro-4-methylpyridine ≥95% for synthesis
2-Amino-5-fluoro-4-methylpyridine

Hazard code(s): Irritant

Formula: C₆H₇FN₂
MW: 126.13
MDL Number: MFCD06659479
CAS Number: 301222-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR