2-Methoxy-4-nitrobenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methoxy-4-nitrobenzene-1-sulphonyl chloride

024778-1G 024778-5G
101818-882EA 27.69 USD
101818-882 101818-884
2-Methoxy-4-nitrobenzenesulfonyl chloride ≥97%
2-Methoxy-4-nitrobenzenesulphonyl chloride

Hazard code(s): Corrosive, Moisture Sensitive

Formula: C₇H₆ClNO₅S
MW: 251.65 g/mol
Melting Pt: 96…98 °C
MDL Number: MFCD03094697
CAS Number: 21320-91-2
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 96-98°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR