1-Boc-4-hydroxypiperidine ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: N-Boc-4-piperidinol, tert-butyl 4-hydroxypiperidine-1-carboxylate, N-BOC-4-Hydroxypiperidine, tert-Butyl 4-hydroxypiperidine-1-carboxylate

024765-25G 024765-100G
101818-852EA 60 USD
101818-852 101818-854
1-Boc-4-hydroxypiperidine ≥98% for synthesis
1-Boc-4-hydroxypiperidine

Hazard code(s): Irritant

Formula: C₁₀H₁₉NO₃
MW: 201.27
Melting Pt: 62…64 °C
MDL Number: MFCD01075174
CAS Number: 109384-19-2

Order Now

Specification Test Results

Melting point 62-64°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR