4-Methoxy-1-butanol ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Methoxybutan-1-ol

024762-5G 024762-25G
101818-842EA 44.62 USD
101818-842 101818-844
4-Methoxy-1-butanol ≥97%
4-Methoxy-1-butanol

Hazard code(s): Irritant, Store in Freezer

Formula: C₅H₁₂O₂
MW: 104.15 g/mol
MDL Number: MFCD00040437
CAS Number: 111-32-0
EINECS: 203-858-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR