(3-Hydroxyphenyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3-Hydroxyphenylacetic acid

024742-25G 024742-100G
101818-790EA 69.23 USD
101818-790 101818-792
(3-Hydroxyphenyl)acetic acid ≥98%
(3-Hydroxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₄(OH))CH₂COOH
MW: 152.15 g/mol
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 128…132 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004337
CAS Number: 621-37-4
EINECS: 210-684-4

Order Now

Specification Test Results

Melting point 128-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR