3-Bromo-6-chloro-2-methylpyridine ≥98%

Supplier: Matrix Scientific

024737-1G 024737-5G
101818-778EA 15.38 USD
101818-778 101818-780
3-Bromo-6-chloro-2-methylpyridine ≥98%
3-Bromo-6-chloro-2-methylpyridine

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₅BrClN
Boiling Pt: 220 °C
Melting Pt: <40 °C
MDL Number: MFCD07375117
CAS Number: 132606-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR