5-Amino-2-methylpyridine ≥98%

Supplier: Matrix Scientific

024730-5G 024730-25G
101818-754EA 47.69 USD
101818-754 102551-068
5-Amino-2-methylpyridine ≥98%
5-Amino-2-methylpyridine

Hazard code(s): Toxic

Formula: C₆H₈N₂
Melting Pt: 95…99 °C
MDL Number: MFCD00833389
CAS Number: 3430-14-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 95-99°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR