2,3-Dichloro-5-nitropyridine ≥98%
Supplier: Matrix Scientific
024729-1G
024729-5G
024729-25G
101818-748EA
15.38
USD
101818-748
101818-750
102663-646
2,3-Dichloro-5-nitropyridine ≥98%
2,3-Dichloro-5-nitropyridine
Hazard code(s): Toxic
Formula:
C₅H₂Cl₂N₂O₂ Melting Pt: 50…52 °C |
MDL Number:
MFCD03840432 CAS Number: 22353-40-8 |
Specification Test Results
Melting point | 50-52°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...