3-Chloro-5-fluoropyridine ≥98%

Supplier: Matrix Scientific

024726-1G 024726-5G
101818-734EA 40 USD
101818-734 101818-736
3-Chloro-5-fluoropyridine ≥98%
3-Chloro-5-fluoropyridine

Hazard code(s): Irritant

Formula: C₅H₃ClFN
Boiling Pt: 126 °C
Density: 1.331 g/cm³
MDL Number: MFCD05663707
CAS Number: 514797-99-0
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR