2-Chloro-4-methyl-5-nitropyridine ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-nitro-4-picoline

024705-5G 024705-25G
101818-690EA 21.54 USD
101818-690 101818-692
2-Chloro-4-methyl-5-nitropyridine ≥98% for synthesis
2-Chloro-4-methyl-5-nitropyridine

Hazard code(s): Irritant

Formula: C₆H₅ClN₂O₂
MW: 172.57 g/mol
MDL Number: MFCD00010688
CAS Number: 23056-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR