4-Bromo-3-chlorobenzoic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
024704-5G
024704-25G
024704-100G
101818-684EA
18.46
USD
101818-684
101818-686
101818-688
4-Bromo-3-chlorobenzoic acid ≥98%
4-Bromo-3-chlorobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₄BrClO₂ Melting Pt: 220…224 °C |
MDL Number:
MFCD08276864 CAS Number: 25118-59-6 |
Specification Test Results
Melting point | 220-224°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...