4-Bromo-2-chlorobenzoic acid ≥98%

Supplier: Matrix Scientific

024703-5G 024703-25G 024703-100G
101818-680EA 21.54 USD
101818-680 101818-682 102663-644
4-Bromo-2-chlorobenzoic acid ≥98%
4-Bromo-2-chlorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Melting Pt: 172…174 °C
MDL Number: MFCD00040903
CAS Number: 59748-90-2
EINECS: 261-911-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 172-174°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR