2-Bromo-1,3,4-thiadiazole ≥98%

Supplier: Matrix Scientific

024702-1G 024702-5G
101818-676EA 44.62 USD
101818-676 101818-678
2-Bromo-1,3,4-thiadiazole ≥98%
2-Bromo-1,3,4-thiadiazole

Hazard code(s): Irritant, Refrigerate, Stored under Argon

Formula: C₂HBrN₂S
Melting Pt: 68…69 °C
MDL Number: MFCD04115375
CAS Number: 61929-24-6

Order Now

Specification Test Results

Melting point 68-69°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR