2-Chloro-6-methoxyisonicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-methoxypyridine-4-carboxylic acid

024693-1G 024693-5G
101818-656EA 76.92 USD
101818-656 101818-658
2-Chloro-6-methoxyisonicotinic acid ≥95%
2-Chloro-6-methoxyisonicotinic acid

Hazard code(s): Irritant

Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
Melting Pt: 210…212 °C
MDL Number: MFCD00173928
CAS Number: 15855-06-8

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR