5-Bromo-2,3-dichloropyridine ≥98%

Supplier: Matrix Scientific

024673-5G 024673-25G
101818-610EA 27.69 USD
101818-610 101818-612
5-Bromo-2,3-dichloropyridine ≥98%
5-Bromo-2,3-dichloropyridine

Hazard code(s): Irritant

Formula: C₅H₂BrCl₂N
MW: 226.89 g/mol
MDL Number: MFCD04112492
CAS Number: 97966-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR