2-(Trifluoromethyl)benzenesulfonamide ≥97%

Supplier: Matrix Scientific

Synonyms: α,α,α-Trifluoro-o-toluenesulphonamide

024671-1G 024671-5G
101818-604EA 15.38 USD
101818-604 101818-606
2-(Trifluoromethyl)benzenesulfonamide ≥97%
2-(Trifluoromethyl)benzenesulphonamide

Hazard code(s): Irritant

Formula: C₇H₆F₃NO₂S
MW: 225.19 g/mol
Melting Pt: 180…184 °C
MDL Number: MFCD00042420
CAS Number: 1869-24-5
UN: 2811
ADR: 6.1,I

Order Now

Specification Test Results

Melting point 180-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR