3-Amino-5-bromo-2-chloropyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-chloro-3-pyridylamine

024664-5G 024664-25G
101818-584EA 23.08 USD
101818-584 101818-586
3-Amino-5-bromo-2-chloropyridine ≥98%
3-Amino-5-bromo-2-chloropyridine

Hazard code(s): Irritant

Formula: C₅H₄BrClN₂
MW: 207.46 g/mol
Melting Pt: 129…132 °C
MDL Number: MFCD02682092
CAS Number: 588729-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR