3-Fluoro-4-nitroaniline ≥97%

Supplier: Matrix Scientific

024653-5G 024653-25G
101818-560EA 81.54 USD
101818-560 101818-562
3-Fluoro-4-nitroaniline ≥97%
3-Fluoro-4-nitroaniline

Hazard code(s): Irritant

Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 152…157 °C (dec.)
MDL Number: MFCD03412239
CAS Number: 2369-13-3
EINECS: 219-130-6

Order Now

Specification Test Results

Melting point 152-157°C(d

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR