5-Bromo-2-chloro-3-fluoropyridine ≥98%

Supplier: Matrix Scientific

024644-1G 024644-5G 024644-25G
101818-538EA 1706.15 USD
101818-538 101818-540 101818-542
5-Bromo-2-chloro-3-fluoropyridine ≥98%
5-Bromo-2-chloro-3-fluoropyridine

Hazard code(s): Irritant

Formula: C₅H₂BrClFN
MDL Number: MFCD03411574
CAS Number: 831203-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR