2-Chloro-6-methyl-5-nitropyridine ≥96% for synthesis
Supplier: Matrix Scientific
Synonyms:
6-Chloro-2-methyl-3-nitropyridine, 2-Chloro-5-nitro-6-picoline, 6-Chloro-3-nitro-2-picoline
024643-5G
024643-25G
101818-534EA
35.38
USD
101818-534
101818-536
2-Chloro-6-methyl-5-nitropyridine ≥96% for synthesis
2-Chloro-6-methyl-5-nitropyridine
Hazard code(s): Irritant
Formula:
C₆H₅ClN₂O₂ |
MDL Number:
MFCD00234160 CAS Number: 22280-60-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...