2-Amino-3-bromo-5-nitropyridine ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 3-Bromo-5-nitro-2-pyridylamine, 3-Bromo-5-nitropyridin-2-amine

024635-5G 024635-25G
101818-508EA 21.54 USD
101818-508 101818-510
2-Amino-3-bromo-5-nitropyridine ≥98% for synthesis
2-Amino-3-bromo-5-nitropyridine

Hazard code(s): Irritant

Formula: C₅H₄BrN₃O₂
MW: 218.01
MDL Number: MFCD00955628
CAS Number: 15862-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR