4'-Fluoro-3'-nitroacetophenone ≥98% for synthesis

Supplier: Matrix Scientific

024634-5G 024634-25G
101818-504EA 21.54 USD
101818-504 101818-506
4'-Fluoro-3'-nitroacetophenone ≥98% for synthesis
4'-Fluoro-3'-nitroacetophenone

Hazard code(s): Irritant

Formula: C₈H₆FNO₃
MW: 183.14
Boiling Pt: 118…120 °C (0.8mm)
Melting Pt: 48…50 °C
MDL Number: MFCD00115369
CAS Number: 400-93-1

Order Now

Specification Test Results

Boiling point 118-120°C/0
Melting point 48-50°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR