2-Fluoro-5-nitro-6-picoline ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Fluoro-2-methyl-3-nitropyridine, 2-Fluoro-6-methyl-5-nitropyridine

024629-1G 024629-5G 024629-25G
101818-490EA 15.38 USD
101818-490 101818-492 101818-494
2-Fluoro-5-nitro-6-picoline ≥98%
2-Fluoro-5-nitro-6-picoline

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₆H₅FN₂O₂
MW: 156.12
MDL Number: MFCD03095071
CAS Number: 18605-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR