2-Fluoro-3-nitro-5-picoline ≥98% for synthesis
Supplier: Matrix Scientific
Synonyms:
2-Fluoro-5-methyl-3-nitropyridine, 2-Fluoro-3-nitro-5-methylpyridine
024625-1G
024625-5G
024625-25G
101818-474EA
27.69
USD
101818-474
101818-476
101818-478
2-Fluoro-3-nitro-5-picoline ≥98% for synthesis
2-Fluoro-3-nitro-5-picoline
Hazard code(s): Irritant, Irritant-Harmful
Formula:
C₆H₅FN₂O₂ MW: 156.12 Boiling Pt: 276 °C Density: 1.357 g/cm³ |
MDL Number:
MFCD09475421 CAS Number: 19346-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...